missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regulatory protein GAL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-76808
This item is not returnable.
View return policy
Description
Regulatory protein GAL4 Polyclonal specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot.
Specifications
| Regulatory protein GAL4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% sodium azide | |
| Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386] | |
| 0.1 mg | |
| Yeast | |
| IgG |
| Western Blot | |
| 1.0 mg/ml | |
| Western Blot 1 - 2 ug/ml | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction