missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Reducing Agent TCEP

Product Code. 12703401
Change view
Click to view available options
:
Reducing Agent TCEP; Volume: 107 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12703401 Reducing Agent TCEP; Volume: 107 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12703401 Supplier Macherey-Nagel™ Supplier No. 740395.107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reducing Agent TCEP is an accessory item for the isolation of RNA and protein.

  • TCEP buffer

Specifications

For Use With (Application) Isolation of RNA and Protein
Content And Storage 15°C to 25°C (Storage)
Quantity 107 mg
compliance-icons
Product Identifier
  • TCEP (107 mg)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: tris-(2-carboxyethyl)phosphine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.