missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Red Silicon Rubber Tubing

Product Code. 10797121 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
10m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10797121 4 mm 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10797121
Only null left
Add to basket
Add to basket

Used in flow lines for fluids and gases in a variety of hydraulic, pneumatic, process.

  • High tear strength, resistance to ozone and ultraviolet radiation performance

Specifications

Diameter (Metric) Inner 4 mm
Material Silicone Rubber
Overall Length (Metric) 10 m
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.