missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rectangular Centrifuge Bucket

Product Code. 10136511 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
5702 Centrifuge rotor A-4-38
Eppendorf rotor A-4-81 (CFA-152-020K)
Swinging bucket rotor CFA-146-010T (A-4-44)
Quantity:
2/Pk.
4/Pk.
Unit Size:
Pack of 2
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity unitSize
10136511 Swinging bucket rotor CFA-146-010T (A-4-44) 4/Pk. Pack of 4
10308612 5702 Centrifuge rotor A-4-38 2/Pk. Pack of 2
10477272 Eppendorf rotor A-4-81 (CFA-152-020K) 4/Pk. Pack of 4
11893882 5702 Centrifuge rotor A-4-38 4/Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10136511 Supplier Eppendorf™ Supplier No. 5804741005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these buckets with Eppendorf™ rotors.

  • Rectangular Centrifuge Buckets
  • Compatible with Eppendorf rotors
  • Available in packs of two and four
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Swinging bucket rotor CFA-146-010T (A-4-44)
Capacity 100 mL
Container Type Bucket
Quantity 4/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.