missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish GH, Mutant Protein
A cDNA sequence encoding the GH, Mutant was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11965-50ug
Additional Details : Weight : 0.01000kg
Specifications
GH, Mutant Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.5% NaHCO3 pH 8. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zebrafish | |
Untagged | |
Greater than 99.0% as determined by: (a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |