missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish GH, Mutant Protein
A cDNA sequence encoding the GH, Mutant was constructed and used to recombinantly synthesize the protein.
£99.15 - £2535.00
Specifications
Name | GH, Mutant Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Zebrafish |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948948
|
enQuireBio™
QP11965-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968948
|
enQuireBio™
QP11965-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958948
|
enQuireBio™
QP11965-1MG |
1 mg |
£2535.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GH, Mutant Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zebrafish | |
Untagged | |
Greater than 99.0% as determined by: (a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.5% NaHCO3 pH 8. |