missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant SARS SARS-CoV Protein

A cDNA sequence encoding the SARS-CoV was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13423-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933519

  • £4295.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SARS SARS-CoV Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASHMSDNG PQSNQRSAPR ITFGGPTDST DNNQNGGRNG ARPKQRRPQG LPNNTASWFT ALTQHGKEEL RFPRGQGVPI NTNSGPDDQI GYYRRATRRV RGGDGKMKEL SPRWYFYYLG TGPEASLPYG ANKEGIVWVA TEGALNTPKD HIGTRNPNNN AATVLQLPQG TTLPKGFYAE GSRGGSQASS RSSSRSRGNS RNSTPGSSRG NSPARMASGG GETALALLLL DRLNQLESKV SGKGQQQQGQ TVTKKSAAEA SKKPRQKRTA TKQYNVTQAF GRRGPEQTQG NFGDQDLIRQ GTDYKHWPQI AQFAPSASAF FGMSRIGMEV TPSGTWLTYH GAIKLDDKDP QFKDNVILLN KHIDAYKTFP PTEPKKDKKK KTDEAQPLPQ RQKKQPTVTL LPAADMDDFS RQLQNSMSGA SADSTQA
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
SARS
His
Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers