missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS-CoV Protein
A cDNA sequence encoding the SARS-CoV was constructed and used to recombinantly synthesize the protein.
£99.15 - £4295.00
Specifications
Name | SARS SARS-CoV Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | SARS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953519
|
enQuireBio™
QP13423-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943519
|
enQuireBio™
QP13423-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933519
|
enQuireBio™
QP13423-1MG |
1 mg |
£4295.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SARS SARS-CoV Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
His | |
Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASHMSDNG PQSNQRSAPR ITFGGPTDST DNNQNGGRNG ARPKQRRPQG LPNNTASWFT ALTQHGKEEL RFPRGQGVPI NTNSGPDDQI GYYRRATRRV RGGDGKMKEL SPRWYFYYLG TGPEASLPYG ANKEGIVWVA TEGALNTPKD HIGTRNPNNN AATVLQLPQG TTLPKGFYAE GSRGGSQASS RSSSRSRGNS RNSTPGSSRG NSPARMASGG GETALALLLL DRLNQLESKV SGKGQQQQGQ TVTKKSAAEA SKKPRQKRTA TKQYNVTQAF GRRGPEQTQG NFGDQDLIRQ GTDYKHWPQI AQFAPSASAF FGMSRIGMEV TPSGTWLTYH GAIKLDDKDP QFKDNVILLN KHIDAYKTFP PTEPKKDKKK KTDEAQPLPQ RQKKQPTVTL LPAADMDDFS RQLQNSMSGA SADSTQA | |
Greater than 95% as determined by SDS-PAGE. |