missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type C-3 Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type C-3, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_000278088.1 |
---|---|
Name | S. aureus Enterotoxin type C-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | entC3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028120
|
enQuireBio™
QP7310-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048120
|
enQuireBio™
QP7310-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038120
|
enQuireBio™
QP7310-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058120
|
enQuireBio™
QP7310-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068120
|
enQuireBio™
QP7310-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078120
|
enQuireBio™
QP7310-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000278088.1 | |
Research Use Only | |
entC3 | |
Recombinant Protein | |
E. coli | |
ESQPDPMPDD LHKSSEFTGT MGNMKYLYDD HYVSATKVKS VDKFLAHDLI YNISDKKLKN YDKVKTELLN EDLAKKYKDE VVDVYGSNYY VNCYFSSKDN VGKVTGGKTC MYGGITKHEG NHFDNGNLQN VLVRVYENKR NTISFEVQTD KKSVTAQELD IKARNFLINK KNLYEFNSSP YETGYIKFIE NNGNTFWYDM MPAPGDKFDQ SKYLMMYNDN KTVDSKSVKI EVHLTTKNG | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Enterotoxin type C-3 Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |