missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rotavirus Non-structural glycoprotein 1

A DNA sequence encoding the Rotavirus A (isolate Monkey/South Africa/SA11-H96/1958 G3-P5B[2]-I2-R2-C5-M5-A5-N5-T5-E2-H5) (RV-A) (Simian Agent 11 (isolate SI/South Africa/H96/58)) Non-structural glycoprotein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7439-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028870

  • £715.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

YP_002302223.1
Rotavirus Non-structural glycoprotein 1
Research Use Only
NSP4
Recombinant Protein
E. coli
PTMKIALKTS KCSYKVVKYC IVTIFNTLLK LAGYKEQITT KDEIEKQMDR VVKEMRRQLE MIDKLTTREI EQVELLKRIY DKLTVQTTGE IDMTKEINQK NVRTLEEWES GKNPYEPREV TAAM
Greater than 90% as determined by SDS-PAGE.
7011361
100 μg
Not determined.
Cytoplasmic Domain
Rotavirus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers