missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rotavirus Non-structural glycoprotein 1
A DNA sequence encoding the Rotavirus A (isolate Monkey/South Africa/SA11-H96/1958 G3-P5B[2]-I2-R2-C5-M5-A5-N5-T5-E2-H5) (RV-A) (Simian Agent 11 (isolate SI/South Africa/H96/58)) Non-structural glycoprotein 4, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | YP_002302223.1 |
---|---|
Gene ID (Entrez) | 7011361 |
Name | Rotavirus Non-structural glycoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028870
|
enQuireBio™
QP7439-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048870
|
enQuireBio™
QP7439-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038870
|
enQuireBio™
QP7439-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058870
|
enQuireBio™
QP7439-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068870
|
enQuireBio™
QP7439-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078870
|
enQuireBio™
QP7439-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
YP_002302223.1 | |
Rotavirus Non-structural glycoprotein 1 | |
Not determined. | |
Cytoplasmic Domain | |
Rotavirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7011361 | |
Research Use Only | |
NSP4 | |
Recombinant Protein | |
E. coli | |
PTMKIALKTS KCSYKVVKYC IVTIFNTLLK LAGYKEQITT KDEIEKQMDR VVKEMRRQLE MIDKLTTREI EQVELLKRIY DKLTVQTTGE IDMTKEINQK NVRTLEEWES GKNPYEPREV TAAM | |
Greater than 90% as determined by SDS-PAGE. |