missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat REG3G / PAP1B Protein
A DNA sequence encoding the Rattus norvegicus (Rat) REG3G / PAP1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6596-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_775120.1 | |
REG3G / PAP1B Protein | |
Research Use Only | |
Reg3g | |
Recombinant Protein | |
Yeast | |
EDAKEDVPTS RISCPKGSRA YGSYCYALFS VSKSWFDADL ACQKRPSGHL VSVLSGSEAS FVSSLIKSSG NSGQNVWIGL HDPTLGQEPN RGGWEWSNAD VMNYFNWETN PSSVSGSHCG TLTRASGFLR WRENNCISEL PYVCKFKA | |
Greater than 90% as determined by SDS-PAGE. |
24620 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |