missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat REG3G / PAP1B Protein
A DNA sequence encoding the Rattus norvegicus (Rat) REG3G / PAP1B, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_775120.1 |
---|---|
Gene ID (Entrez) | 24620 |
Name | REG3G / PAP1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004430
|
enQuireBio™
QP6596-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044430
|
enQuireBio™
QP6596-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014430
|
enQuireBio™
QP6596-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034430
|
enQuireBio™
QP6596-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054430
|
enQuireBio™
QP6596-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024430
|
enQuireBio™
QP6596-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_775120.1 | |
REG3G / PAP1B Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24620 | |
Research Use Only | |
Reg3g | |
Recombinant Protein | |
Yeast | |
EDAKEDVPTS RISCPKGSRA YGSYCYALFS VSKSWFDADL ACQKRPSGHL VSVLSGSEAS FVSSLIKSSG NSGQNVWIGL HDPTLGQEPN RGGWEWSNAD VMNYFNWETN PSSVSGSHCG TLTRASGFLR WRENNCISEL PYVCKFKA | |
Greater than 90% as determined by SDS-PAGE. |