missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PVALB Protein
A cDNA sequence encoding the PVALB was constructed and used to recombinantly synthesize the protein.
£99.15 - £2645.00
Specifications
Gene ID (Entrez) | 25269 |
---|---|
Name | PVALB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Pvalb |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15982879
|
enQuireBio™
QP13219-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15972879
|
enQuireBio™
QP13219-10UG |
10 μg |
£302.00
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962879
|
enQuireBio™
QP13219-100UG |
100 μg |
£2645.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
25269 | |
Research Use Only | |
Pvalb | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) without any additives. |
PVALB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSMTDLLSAE DIKKAIGAFT AADSFDHKKF FQMVGLKKKS ADDVKKVFHI LDKDKSGFIE EDELGSILKG FSSDARDLSA KETKTLMAAG DKDGDGKIGV EEFSTLVAES | |
Greater than 90% as determined by SDS-PAGE. |