missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PVALB Protein
A cDNA sequence encoding the PVALB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13219-10ug
Additional Details : Weight : 0.01000kg
Specifications
25269 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSMTDLLSAE DIKKAIGAFT AADSFDHKKF FQMVGLKKKS ADDVKKVFHI LDKDKSGFIE EDELGSILKG FSSDARDLSA KETKTLMAAG DKDGDGKIGV EEFSTLVAES | |
Greater than 90% as determined by SDS-PAGE. |
PVALB Protein | |
Research Use Only | |
Pvalb | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) without any additives. |