missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLBD2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLBD2, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_640348.2 |
---|---|
Gene ID (Entrez) | 246120 |
Name | PLBD2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010051
|
enQuireBio™
QP7660-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050051
|
enQuireBio™
QP7660-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020051
|
enQuireBio™
QP7660-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040051
|
enQuireBio™
QP7660-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060051
|
enQuireBio™
QP7660-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030051
|
enQuireBio™
QP7660-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_640348.2 | |
PLBD2 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
246120 | |
Research Use Only | |
Plbd2 | |
Recombinant Protein | |
Yeast | |
GALPTLGPGW RRQNPEPPAS RTRSLLLDAA SGQLRLEYGF HPDAVAWANL TNAIRETGWA YLDLGTNGSY NDSLQAYAAG VVEASVSEEL IYMHWMNTVV NYCGPFEYEV GYCEKLKSFL EANLEWMQRE MELSPDSPYW HQVRLTLLQL KGLEDSYEGR LTFPTGRFNI KPLGFLLLQI SGDLEDLEPA LNKTNTKPSV GSGSCSALIK LLPGSHDLLV AHNTWNSYQN MLRIIKKYRL QFREGPQEEY PLIAGNNLIF SSYPGTIFSG DDFYILGSGL VTLETTIGNK NPALWKYVQP QGCVLEWIRN IVANRLALDG ATWADVFRRF NSGTYNNQWM IVDYKAFIPN GPSPGSRVLT ILEQIPGMVV VADKTAELYK TTYWASYNIP YFESVFNASG LQALVAQYGD WFSYTRNPRA KIFQRDQSLV EDVDTMVRLM RYNDFLHDPL SLCEACSPKP NAENAISARS DLNPANGSYP FQALRQRAHG GIDVKVTSVA LAKYMSMLAA SGPTWDQLPP FQWSKSPFHN MLHMGQPDLW MFSPVKVPWD | |
Greater than 90% as determined by SDS-PAGE. |