missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL17 / IL17a Protein
Recombinant Protein
Brand: enQuireBio™ QP5511-25ug
Additional Details : Weight : 0.01000kg
Specifications
IL17 / IL17a Protein | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS | |
Greater than 95% as determined by SDS-PAGE. |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL17 / IL17a induces IL-6 production in 3T3 cells measured via ELISA (typical ED50 is ? 1 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium citrate, pH 3.0 |