missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL17 / IL17a Protein
Recombinant Protein
£80.80 - £4295.00
Specifications
Name | IL17 / IL17a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | IL17A |
Biological Activity | Rat IL17 / IL17a induces IL-6 production in 3T3 cells measured via ELISA (typical ED50 is ? 1 ng/ml). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906299
|
enQuireBio™
QP5511-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986289
|
enQuireBio™
QP5511-25UG |
25 μg |
£191.00
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966289
|
enQuireBio™
QP5511-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996289
|
enQuireBio™
QP5511-500UG |
500 μg |
£2625.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976289
|
enQuireBio™
QP5511-1MG |
1 mg |
£4295.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL17 / IL17a Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL17 / IL17a induces IL-6 production in 3T3 cells measured via ELISA (typical ED50 is ? 1 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium citrate, pH 3.0 |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS | |
Greater than 95% as determined by SDS-PAGE. |