Learn More
enQuireBio™ Recombinant Rat IL17 / IL17a Protein
Recombinant Protein
Brand:  enQuireBio™ QP5511-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15966289
  • £590.00 / 100µg
Estimated Shipment: 20-09-2022
to see stock.
Specifications
IL17 / IL17a Protein
100 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Rat IL17 / IL17a induces IL-6 production in 3T3 cells measured via ELISA (typical ED50 is ? 1 ng/ml).
Rat
Untagged
10 mM sodium citrate, pH 3.0
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL17A
Recombinant Protein
E. coli
MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS