missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 21 Protein

A cDNA sequence encoding the IL 21 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10702-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954908

  • £155.00 / 10µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

365769
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The biological activity is measured by its ability to proliferate activated B cells.
Rat
Untagged
Rat IL-21 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
IL 21 Protein
Research Use Only
Il21
Recombinant Protein
E. coli
HKSSPQRPDH LLIRLRHLMD IVEQLKIYEN DLDPELLTAP QDVKGQCEHE AFACFQKAKL KPSNTGNNKT FINDLLAQLR RRLPAKRTGN KQRHMAKCPS CDLYEKKTPK EFLERLKWLL QKMIHQHLS
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers