missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 21 Protein
A cDNA sequence encoding the IL 21 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5160.00
Specifications
Gene ID (Entrez) | 365769 |
---|---|
Name | IL 21 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974908
|
enQuireBio™
QP10702-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954908
|
enQuireBio™
QP10702-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964908
|
enQuireBio™
QP10702-1MG |
1 mg |
£5160.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
365769 | |
Research Use Only | |
Il21 | |
Recombinant Protein | |
E. coli | |
HKSSPQRPDH LLIRLRHLMD IVEQLKIYEN DLDPELLTAP QDVKGQCEHE AFACFQKAKL KPSNTGNNKT FINDLLAQLR RRLPAKRTGN KQRHMAKCPS CDLYEKKTPK EFLERLKWLL QKMIHQHLS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 21 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity is measured by its ability to proliferate activated B cells. | |
Rat | |
Untagged | |
Rat IL-21 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |