missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 13 Protein
A cDNA sequence encoding the IL 13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10692-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
116553 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
ED50 range = 40ng/ml, corresponding to a specific activity of > 25,000IU/mg as determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay. | |
Rat | |
Untagged | |
The protein (1 mg/ml) was lyophilized in PBS, pH 7.4. |
IL 13 Protein | |
Research Use Only | |
Il13 | |
Recombinant Protein | |
E. coli | |
VRRSTSPPVA LRELIEELSN ITQDQKTSLC NSSIVWSVDI TAGGFCAALE SLTNISSCNA IHRTQRILNG LCNQKASDVA SSPPDTKIEV AQFISKLLNY SKQLFRYGH | |
Greater than 95% as determined by SDS-PAGE. |