missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 13 Protein
A cDNA sequence encoding the IL 13 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5160.00
Specifications
Gene ID (Entrez) | 116553 |
---|---|
Name | IL 13 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il13 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974668
|
enQuireBio™
QP10692-EC-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954668
|
enQuireBio™
QP10692-EC-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964668
|
enQuireBio™
QP10692-EC-1MG |
1 mg |
£5160.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
116553 | |
Research Use Only | |
Il13 | |
Recombinant Protein | |
E. coli | |
VRRSTSPPVA LRELIEELSN ITQDQKTSLC NSSIVWSVDI TAGGFCAALE SLTNISSCNA IHRTQRILNG LCNQKASDVA SSPPDTKIEV AQFISKLLNY SKQLFRYGH | |
Greater than 95% as determined by SDS-PAGE. |
IL 13 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
ED50 range = 40ng/ml, corresponding to a specific activity of > 25,000IU/mg as determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay. | |
Rat | |
Untagged | |
The protein (1 mg/ml) was lyophilized in PBS, pH 7.4. |