missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF9 Protein
Recombinant Protein
Brand: enQuireBio™ QP5514-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037084.1 | |
FGF9 Protein | |
Research Use Only | |
FGF9 | |
Recombinant Protein | |
E. coli | |
MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS | |
Greater than 95% as determined by SDS-PAGE. |
25444 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat FGF9 induces THP-1 chemotaxis (significanty different from control at Rat FGF9 concentrations greater than 5 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, 75 mM ammonium sulfate, pH 7.5 |