missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF9 Protein
Recombinant Protein
£80.80 - £4075.00
Specifications
Accession Number | NP_037084.1 |
---|---|
Gene ID (Entrez) | 25444 |
Name | FGF9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946309
|
enQuireBio™
QP5514-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926309
|
enQuireBio™
QP5514-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916309
|
enQuireBio™
QP5514-100UG |
100 μg |
£535.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956309
|
enQuireBio™
QP5514-500UG |
500 μg |
£2495.00
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936309
|
enQuireBio™
QP5514-1MG |
1 mg |
£4075.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_037084.1 | |
FGF9 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat FGF9 induces THP-1 chemotaxis (significanty different from control at Rat FGF9 concentrations greater than 5 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, 75 mM ammonium sulfate, pH 7.5 |
25444 | |
Research Use Only | |
FGF9 | |
Recombinant Protein | |
E. coli | |
MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS | |
Greater than 95% as determined by SDS-PAGE. |