missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Cyclophilin A Protein

A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11578-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967698

  • £99.15 / 2µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

100360977; 25518
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKHHHHHHASVNPTVFFDIT ADGEPLGRVC FELFADKVPK TAENFRALST GEKGFGYKGS SFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMSIVEA MERFGSRNGK TSKKITISDC GQL
Greater than 95.0% as determined by SDS-PAGE.
Cyclophilin A Protein
Research Use Only
Ppia
Rat
His
Cyclophilin-A was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline and 5%(w/v) trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers