missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cyclophilin A Protein
A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 100360977; 25518 |
---|---|
Name | Cyclophilin A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Ppia |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967698
|
enQuireBio™
QP11578-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947698
|
enQuireBio™
QP11578-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957698
|
enQuireBio™
QP11578-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100360977; 25518 | |
Research Use Only | |
Ppia | |
Rat | |
His | |
Cyclophilin-A was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline and 5%(w/v) trehalose. |
Cyclophilin A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVNPTVFFDIT ADGEPLGRVC FELFADKVPK TAENFRALST GEKGFGYKGS SFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMSIVEA MERFGSRNGK TSKKITISDC GQL | |
Greater than 95.0% as determined by SDS-PAGE. |