missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL5 / RANTES Protein
Recombinant Protein
Brand: enQuireBio™ QP5513-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038681.2 | |
CCL5 / RANTES Protein | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS | |
Greater than 95% as determined by SDS-PAGE. |
20304 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 250 ng/ml of Rat CCL5 / RANTES. | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |