missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL5 / RANTES Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_038681.2 |
---|---|
Gene ID (Entrez) | 20304 |
Name | CCL5 / RANTES Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906309
|
enQuireBio™
QP5513-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986299
|
enQuireBio™
QP5513-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966299
|
enQuireBio™
QP5513-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996299
|
enQuireBio™
QP5513-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976299
|
enQuireBio™
QP5513-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_038681.2 | |
CCL5 / RANTES Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 250 ng/ml of Rat CCL5 / RANTES. | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
20304 | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS | |
Greater than 95% as determined by SDS-PAGE. |