missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL2 / MCP-1 / MCP1 Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_113718.1 |
---|---|
Gene ID (Entrez) | 24770 |
Name | CCL2 / MCP-1 / MCP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946289
|
enQuireBio™
QP5509-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926289
|
enQuireBio™
QP5509-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916289
|
enQuireBio™
QP5509-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956289
|
enQuireBio™
QP5509-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936289
|
enQuireBio™
QP5509-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_113718.1 | |
CCL2 / MCP-1 / MCP1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat CCL2 / MCP-1 / MCP1 induces THP-1 chemotaxis (significanty different from control at Rat CCL2 / MCP-1 / MCP1 concentrations greater than 10 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
24770 | |
Research Use Only | |
CCL2 | |
Recombinant Protein | |
E. coli | |
QPDAVNAPLT CCYSFTGKMI PMSRLENYKR ITSSRCPKEA VVFVTKLKRE ICADPNKEWV QKYIRKLDQN QVRSETTVFY KIASTLRTSA PLNVNLTHKS EANASTLFST TTSSTSVEVT SMTEN | |
Greater than 95% as determined by SDS-PAGE. |