missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CCL2 / MCP-1 / MCP1 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5509-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936289

  • £3085.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_113718.1
CCL2 / MCP-1 / MCP1 Protein
Research Use Only
CCL2
Recombinant Protein
E. coli
QPDAVNAPLT CCYSFTGKMI PMSRLENYKR ITSSRCPKEA VVFVTKLKRE ICADPNKEWV QKYIRKLDQN QVRSETTVFY KIASTLRTSA PLNVNLTHKS EANASTLFST TTSSTSVEVT SMTEN
Greater than 95% as determined by SDS-PAGE.
24770
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Rat CCL2 / MCP-1 / MCP1 induces THP-1 chemotaxis (significanty different from control at Rat CCL2 / MCP-1 / MCP1 concentrations greater than 10 ng/ml).
Rat
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers