missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Calcium-dependent phospholipase A2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6500-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013760

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_058870.1
Calcium-dependent phospholipase A2 Protein
Research Use Only
PLA2G5
Recombinant Protein
E. coli
GLLELKSMIE KVTGKNAVKN YGFYGCYCGW GGHGTPKDGT DWCCRMHDRC YGLLEEKHCA IRTQSYDYRF TQDLVICEHD SFCPVRLCAC DRKLVYCLRR NLWSYNRLYQ YYPNFLC
Greater than 90% as determined by SDS-PAGE.
29354
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers