missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Calcium-dependent phospholipase A2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_058870.1 |
---|---|
Gene ID (Entrez) | 29354 |
Name | Calcium-dependent phospholipase A2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083750
|
enQuireBio™
QP6500-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003760
|
enQuireBio™
QP6500-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093750
|
enQuireBio™
QP6500-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013760
|
enQuireBio™
QP6500-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023760
|
enQuireBio™
QP6500-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033760
|
enQuireBio™
QP6500-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058870.1 | |
Calcium-dependent phospholipase A2 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29354 | |
Research Use Only | |
PLA2G5 | |
Recombinant Protein | |
E. coli | |
GLLELKSMIE KVTGKNAVKN YGFYGCYCGW GGHGTPKDGT DWCCRMHDRC YGLLEEKHCA IRTQSYDYRF TQDLVICEHD SFCPVRLCAC DRKLVYCLRR NLWSYNRLYQ YYPNFLC | |
Greater than 90% as determined by SDS-PAGE. |