missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit CXCL9 / MIG / C-X-C motif chemokine 9 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) CXCL9 / MIG / C-X-C motif chemokine 9, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Name | CXCL9 / MIG / C-X-C motif chemokine 9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CXCL9 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958519
|
enQuireBio™
QP5897-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978519
|
enQuireBio™
QP5897-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968519
|
enQuireBio™
QP5897-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988519
|
enQuireBio™
QP5897-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998519
|
enQuireBio™
QP5897-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908529
|
enQuireBio™
QP5897-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CXCL9 / MIG / C-X-C motif chemokine 9 Protein | |
Not determined. | |
Full Length | |
Rabbit | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
CXCL9 | |
Recombinant Protein | |
E. coli | |
SPIMRNGRCS CISSTQGKIH LQSLKDLKQF SPSPSCGKTE IIATKKDGTQ ICLNPDSTEV KELVEKWKKQ SSPKKKQKKG KKQRKVKKSL KKSQRPHQKK TA | |
Greater than 90% as determined by SDS-PAGE. |