missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit ALDOB / Aldolase B Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) ALDOB / Aldolase B, was expressed in the hosts and tags indicated.

£321.05 - £2965.00

Specifications

Accession Number NP_001164585.1
Gene ID (Entrez) 100328925
Name ALDOB / Aldolase B Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15946699
View Documents Promotion Details
enQuireBio™
QP5646-YE-100UG
100 μg
£800.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15966699
View Documents Promotion Details
enQuireBio™
QP5646-YE-1MG
1 mg
£2965.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15956699
View Documents Promotion Details
enQuireBio™
QP5646-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15976699
View Documents Promotion Details
enQuireBio™
QP5646-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15986699
View Documents Promotion Details
enQuireBio™
QP5646-YE-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15996699
View Documents Promotion Details
enQuireBio™
QP5646-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001164585.1
ALDOB / Aldolase B Protein
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
100328925
Research Use Only
ALDOB
Recombinant Protein
Yeast
AHRFPALTPE QKKELSDIAQ RIVANGKGIL AADESVGTMG NRLQRIKVEN TEENRRQFRE ILFTVDNSIN QSIGGVILFH ETLYQKDSQG KLFRNILKEK GIVVGIKLDQ GGAPLAGTNK ETTIQGLDGL SERCAQYKKD GVDFGKWRAV LRIADQCPSS LAIQENANTL ARYASICQQN GLVPIVEPEV IPDGDHDLEH CQYVTEKVLA AVYKALNDHH VYLEGTLLKP NMVTAGHACT KKYTPEQVAM ATVTALHRTV PAAVPGICFL SGGMSEEDAT LNLNAINLCP LPKPWKLSFS YGRALQASAL AAWGGKAENK KATQEAFMKR AVVNCQAAKG QYVHTGSSGA ASTQSLFTAS YTY
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications