missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit ALDOB / Aldolase B Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) ALDOB / Aldolase B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5646-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966699

  • £2965.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001164585.1
ALDOB / Aldolase B Protein
Research Use Only
ALDOB
Recombinant Protein
Yeast
AHRFPALTPE QKKELSDIAQ RIVANGKGIL AADESVGTMG NRLQRIKVEN TEENRRQFRE ILFTVDNSIN QSIGGVILFH ETLYQKDSQG KLFRNILKEK GIVVGIKLDQ GGAPLAGTNK ETTIQGLDGL SERCAQYKKD GVDFGKWRAV LRIADQCPSS LAIQENANTL ARYASICQQN GLVPIVEPEV IPDGDHDLEH CQYVTEKVLA AVYKALNDHH VYLEGTLLKP NMVTAGHACT KKYTPEQVAM ATVTALHRTV PAAVPGICFL SGGMSEEDAT LNLNAINLCP LPKPWKLSFS YGRALQASAL AAWGGKAENK KATQEAFMKR AVVNCQAAKG QYVHTGSSGA ASTQSLFTAS YTY
Greater than 90% as determined by SDS-PAGE.
100328925
1 mg
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers