missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine UMOD Protein
A cDNA sequence encoding the UMOD was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | UMOD Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Porcine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955039
|
enQuireBio™
QP13892-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935039
|
enQuireBio™
QP13892-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945039
|
enQuireBio™
QP13892-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
UMOD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
The UMOD protein was lyophilized from 0.4microm filtered solution at a concentration of 0.5 mg/ml containing deionized water. |
Research Use Only | |
Recombinant Protein | |
Porcine | |
MRWLFSLPSI WMVVAVTSWI IMAAAEDTSE ARSCSECHSN ATCVEDGAAT TCTCRVGFTG DGLECVDLDE CSLPGAHDCS ANSSCVNTPG SYTCVCSDGF RLSPELGCID VDECAEPGLS RCHALATCVN GEGNYSCVCP AGYWGDGWHC ECSPGSCGPG LDCVPSGDAL MCADPCQAHR ILDEYWRSAG YGAGYDCDSD LSGWYRFVGQ GGVRLADTCV PVLRCNTAAP MWLNGTHPSS DEGIVDRTAC AHWSGHCCLW NTPVQVKACD GYYVYNLTAP PECHLAYCTD PSSVEGTCEE CSIDEDCKSD NGKWHCQCKQ DVNITDFSLL EHRLECGVND IKMSLSKCQL KNLGFEKIFM YLRDSQCSGF AERGNRDWIS VVTPTRDGPC GTVMTRNETH ATYSNTLYLA DEIIIRDLNI KINFACSYPL DMKVSLKTSL QPMVSALNIS VGGTGMFTVR MALFQSPTYT QPYQGSSVTL STEAFLFVGT MLDGGDLSRF VLLMTNCYAT PSSSATDPLK YFIIQDRCPR SQDPTIQVEE NGESPQGRFS VQMFRFAGNY DLVYLHCEVY LCDSVNEKCK PTCSGTRFRS GGALDQSRVL NLGPITRKGG QAAVSRAASS SLGFLKVWLP LLLSATLTLM SQ |