missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine UMOD Protein

A cDNA sequence encoding the UMOD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13892-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945039

  • £5730.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UMOD Protein
Research Use Only
Recombinant Protein
Porcine
MRWLFSLPSI WMVVAVTSWI IMAAAEDTSE ARSCSECHSN ATCVEDGAAT TCTCRVGFTG DGLECVDLDE CSLPGAHDCS ANSSCVNTPG SYTCVCSDGF RLSPELGCID VDECAEPGLS RCHALATCVN GEGNYSCVCP AGYWGDGWHC ECSPGSCGPG LDCVPSGDAL MCADPCQAHR ILDEYWRSAG YGAGYDCDSD LSGWYRFVGQ GGVRLADTCV PVLRCNTAAP MWLNGTHPSS DEGIVDRTAC AHWSGHCCLW NTPVQVKACD GYYVYNLTAP PECHLAYCTD PSSVEGTCEE CSIDEDCKSD NGKWHCQCKQ DVNITDFSLL EHRLECGVND IKMSLSKCQL KNLGFEKIFM YLRDSQCSGF AERGNRDWIS VVTPTRDGPC GTVMTRNETH ATYSNTLYLA DEIIIRDLNI KINFACSYPL DMKVSLKTSL QPMVSALNIS VGGTGMFTVR MALFQSPTYT QPYQGSSVTL STEAFLFVGT MLDGGDLSRF VLLMTNCYAT PSSSATDPLK YFIIQDRCPR SQDPTIQVEE NGESPQGRFS VQMFRFAGNY DLVYLHCEVY LCDSVNEKCK PTCSGTRFRS GGALDQSRVL NLGPITRKGG QAAVSRAASS SLGFLKVWLP LLLSATLTLM SQ
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Porcine
Untagged
The UMOD protein was lyophilized from 0.4microm filtered solution at a concentration of 0.5 mg/ml containing deionized water.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers