missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine LH Protein

A cDNA sequence encoding the LH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12568-100IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940839

  • £99.15 / 100 units
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

397153
100 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Porcine
MEMLQGLLLW LLLSVAGVWA SRGPLRPLCR PINATLAAEN EACPVCITFT TSICAGYCPS MVRVLPAALP PVPQPVCTYR ELSFASIRLP GCPPGVDPTV SFPVALSCHC GPCRLSSSDC GGPRAQPLAC DRPLLPGLLF L
LH Protein
Research Use Only
LHB
Porcine
Untagged
The LH was lyophilized with no additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers