missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine LH Protein
A cDNA sequence encoding the LH was constructed and used to recombinantly synthesize the protein.
£99.15 - £770.00
Specifications
Gene ID (Entrez) | 397153 |
---|---|
Name | LH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LHB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930839
|
enQuireBio™
QP12568-10,000IU |
10000 IU |
£770.00
10000 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940839
|
enQuireBio™
QP12568-100IU |
100 IU |
£99.15
100 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950839
|
enQuireBio™
QP12568-750IU |
750 IU |
£155.00
750 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
397153 | |
Research Use Only | |
LHB | |
Porcine | |
Untagged | |
The LH was lyophilized with no additives. |
LH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Porcine | |
MEMLQGLLLW LLLSVAGVWA SRGPLRPLCR PINATLAAEN EACPVCITFT TSICAGYCPS MVRVLPAALP PVPQPVCTYR ELSFASIRLP GCPPGVDPTV SFPVALSCHC GPCRLSSSDC GGPRAQPLAC DRPLLPGLLF L |