missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TrxR Yeast Protein
A cDNA sequence encoding the TrxR Yeast was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10895-5ug
Additional Details : Weight : 0.01000kg
Specifications
TrxR Yeast Protein | |
Research Use Only | |
The specific activity was found to be 5.8 IU/mg. | |
Other | |
Untagged | |
Each mg of protein contains 20mM phosphate buffer pH 7.4. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKNLTNEIYD VIIVGSGPAG LTSAIYTARA GLKTVIYEQS APGGKLIKTD LIENYPGFET IQGPDLASKM YLQALSLNAS VEFVGVDSIF KNADDIFEIS LTNGEIKKAY SVILATGTQE NKLGIKGEDE LYGKGVSYCA VCDGAFYKNK PVAVVGGGYS AVQEAMYLSQ LVDKVYLIVR RDVFRADANK VAKLKAQPNV EFLLKSQVKE IHGAEKVESL TITTPDGEKK LVVSAIFPYI GSTPLIDSVK HLCIENEKGY IPTNDKMQTE IKGLFVAGDV RDVPLRQIAI ACGDGAIAGQ MAVEYVQEVK | |
Greater than 95.0% as determined by SDS-PAGE. |