missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TrxR Yeast Protein
A cDNA sequence encoding the TrxR Yeast was constructed and used to recombinantly synthesize the protein.
£99.15 - £3525.00
Specifications
Name | TrxR Yeast Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity was found to be 5.8 IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945498
|
enQuireBio™
QP10895-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935498
|
enQuireBio™
QP10895-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925498
|
enQuireBio™
QP10895-1MG |
1 mg |
£3525.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TrxR Yeast Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKNLTNEIYD VIIVGSGPAG LTSAIYTARA GLKTVIYEQS APGGKLIKTD LIENYPGFET IQGPDLASKM YLQALSLNAS VEFVGVDSIF KNADDIFEIS LTNGEIKKAY SVILATGTQE NKLGIKGEDE LYGKGVSYCA VCDGAFYKNK PVAVVGGGYS AVQEAMYLSQ LVDKVYLIVR RDVFRADANK VAKLKAQPNV EFLLKSQVKE IHGAEKVESL TITTPDGEKK LVVSAIFPYI GSTPLIDSVK HLCIENEKGY IPTNDKMQTE IKGLFVAGDV RDVPLRQIAI ACGDGAIAGQ MAVEYVQEVK | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
The specific activity was found to be 5.8 IU/mg. | |
Other | |
Untagged | |
Each mg of protein contains 20mM phosphate buffer pH 7.4. |