missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other scIC2 Protein
A cDNA sequence encoding the scIC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13433-1mg
Additional Details : Weight : 0.01000kg
Specifications
scIC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein solution contains 20mM Tris-HCl, 250mM NaCl and 50% glycerol, pH 7.5. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 91.0% as determined by SDS-PAGE. |