missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other scIC2 Protein
A cDNA sequence encoding the scIC2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | scIC2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953549
|
enQuireBio™
QP13433-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933549
|
enQuireBio™
QP13433-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943549
|
enQuireBio™
QP13433-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
scIC2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 91.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein solution contains 20mM Tris-HCl, 250mM NaCl and 50% glycerol, pH 7.5. |