missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MIF T. Vaginalis Protein
A cDNA sequence encoding the MIF T. Vaginalis was constructed and used to recombinantly synthesize the protein.
£104.00 - £6030.00
Specifications
Name | MIF T. Vaginalis Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | E. coli |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15981259
|
enQuireBio™
QP12697-2UG |
2 μg |
£104.00
2µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15961259
|
enQuireBio™
QP12697-10UG |
10 μg |
£159.00
10µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15971259
|
enQuireBio™
QP12697-1MG |
1 mg |
£6030.00
1mg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
MIF T. Vaginalis Protein | |
His | |
Other | |
Greater than 95.0% as determined by SDS-PAGE. |
Recombinant Protein | |
E. coli | |
MPALVIKTNA KFTEEEKSKA TEELGNIVSK VLGKPISYVM VTLEDGVAVR FGGSDEKAAF MSLMSIGGLN RAVNKRASAA LTKWFTDHGF QGDRIYIVFN PKSAEDWGFN GDTFAHHHHH H |