missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MIF T. Vaginalis Protein
A cDNA sequence encoding the MIF T. Vaginalis was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12697-1mg
Additional Details : Weight : 0.01000kg
Specifications
MIF T. Vaginalis Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPALVIKTNA KFTEEEKSKA TEELGNIVSK VLGKPISYVM VTLEDGVAVR FGGSDEKAAF MSLMSIGGLN RAVNKRASAA LTKWFTDHGF QGDRIYIVFN PKSAEDWGFN GDTFAHHHHH H | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
MIF T. Vaginalis filtered (0.4 microm) and lyophilized from 0.5 mg/ml solution in 20 mM Tris buffer and 50 mM NaCl, pH 7.5. |