missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other L-Asparaginase Protein
A cDNA sequence encoding the L-Asparaginase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10740-500IU
Additional Details : Weight : 0.01000kg
Specifications
L-Asparaginase Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The enzyme was lyophilized with no additives. |
500 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Other | |
Untagged | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |