missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other L-Asparaginase Protein
A cDNA sequence encoding the L-Asparaginase was constructed and used to recombinantly synthesize the protein.
£99.15 - £439.00
Specifications
Name | L-Asparaginase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15925028
|
enQuireBio™
QP10740-10000IU |
10000 IU |
£439.00
10000 units |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935028
|
enQuireBio™
QP10740-2500IU |
2500 IU |
£160.00
2500 units |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945028
|
enQuireBio™
QP10740-500IU |
500 IU |
£99.15
500 units |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
L-Asparaginase Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Other | |
Untagged | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The enzyme was lyophilized with no additives. |