missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other L-Asparaginase Protein

A cDNA sequence encoding the L-Asparaginase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10740-2500IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935028

  • £160.00 / 2500 units
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

L-Asparaginase Protein
Research Use Only
Recombinant Protein
E. coli
The enzyme was lyophilized with no additives.
2500 IU
< 1.0 EU per ug protein as determined by the LAL method.
Other
Untagged
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers