missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Dnak SBD C-terminus Protein

A cDNA sequence encoding the Dnak SBD C-terminus was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11688-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928048

  • £155.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dnak SBD C-terminus Protein
Research Use Only
Recombinant Protein
E. coli
MDVKDVLLLD VTPLSLGIET MGGVMTTLIA KNTTIPTKHS QVFSTAEDNQ SAVTIHVLQGERKRAADNKS LGQFNLDGIN PAPRGMPQIE VTFDIDADGI LHVSAKDKNS GKEQKITIKA SSGLNEDEIQ KMVRDAEANA EADRKFEELV QTRNQGDHLL HSTRKQVEEA GDKLPADDKTAIESALTALE TALKGEDKAA IEAKMQELAQ VSQKLMEIAQ QQHAQQQTAG ADASANNAKD DDVVDAEFEE VKDKK
Greater than 95.0% as determined by SDS-PAGE.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The DnaK protein contains 25mM Tris-HCl, pH 7.5, 100mM NaCl, 5mM DTT and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers